White spirit, 220 …300 °C Stoddard Solvent

Supplier: AQUA SOLUTIONS MS
Danger

S6010-4L
101226-376EA 169.1 USD
101226-376
White spirit, 220 …300 °C Stoddard Solvent
White Spirit
Flash Pt: 40 °C
CAS Number: 8052-41-3
UN: 1268
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR