Hydrogen peroxide solution 31% (w/w)

Supplier: AQUA SOLUTIONS MS

H3000-100ML
101226-170EA 37.07 USD
101226-170
Hydrogen peroxide solution 31% (w/w)
Hydrogen peroxide
Formula: H₂O₂
MDL Number: MFCD00011333
CAS Number: 7722-84-1
UN: 2014
ADR: 5.1,II

Order Now

Specification Test Results

Aqueous solutions 20 - 40%
Case Size 12 × 100 mL


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR