Sodium hydroxide 0.1 M traceable to NIST Standard Reference Material (SRM)
Supplier: AQUA SOLUTIONS MS
|
Danger
|
Synonyms:
Caustic soda, Caustic soda, Sodium hydrate, Lye, Soda lye
8625-20L
8625-4LC
8625-1L
8625-10L
8625-500ML
8625-4L
101225-214EA
162.59
USD
101225-214
101223-014
101223-036
101223-190
101223-406
101223-140
Sodium hydroxide 0.1 M traceable to NIST Standard Reference Material (SRM)
Sodium hydroxide
Formula:
NaOH |
MDL Number:
MFCD00003548 CAS Number: 1310-73-2 |
Specification Test Results
Normality | 0.1000 ± 0.0005 Normal @ 20° C |
Appearance and Color | Clear Water White Liquid |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...