Sulfuric acid 50% (v/v) 18 N
Supplier: AQUA SOLUTIONS MS
|
Danger
|
9000-1L
9000-20L
9000-500ML
9000-4L
9000-500MLG
101223-108EA
48.03
USD
101223-108
101225-186
101226-548
101225-318
103193-862
Sulfuric acid 50% (v/v) 18 N
Sulfuric acid
Formula:
H₂SO₄ MW: 98.08 Boiling Pt: 330 Melting Pt: 10.38 Density: 1.42 g/cm³ (11.85 lbs/gal.) (20 °C) |
MDL Number:
MFCD00064589 CAS Number: 7664-93-9 UN: 2796 |
Specification Test Results
Assay | 50% |
Normality | 18 |
Appearance | Clear Liquid |
Vapor Pressure (20 °C) | 23 hPa (17 mm Hg) |
Density (20 °C) | 1.48231 - 1.50189 |
Odor | Odorless |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...