1,2-Dichloroethane ≥99.0% ACS

Supplier: AQUA SOLUTIONS MS
Danger

D4490-20L D4490-4L D4490-1L D4490-500ML
101225-076EA 726.86 USD
101225-076 101224-992 101224-990 101226-114
1,2-Dichloroethane ≥99.0% ACS
1,2-Dichloroethane
Formula: ClH₂CCH₂Cl
MDL Number: MFCD00000963
CAS Number: 107-06-2
EINECS: 203-458-1
UN: 1184
ADR: 3,6.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=48789e88e6041156e33712fd7c0198d7a4910266&vid=4238623c54017660b83d3baa856a69cc1aa8ee6d&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR