Starch indicator 2% (w/v) in aqueous solution

Supplier: AQUA SOLUTIONS MS

8890-500ML 8890-1L
101224-584EA 62.87 USD
101224-584 101225-594
Starch indicator 2% (w/v) in aqueous solution
Starch
Formula: (C₆H₁₀O₅)n
MDL Number: MFCD00082026
CAS Number: 9005-25-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR