Glycerine ≥99.5%

Supplier: AQUA SOLUTIONS MS

G7502-4L
101224-392EA 279.14 USD
101224-392
Glycerine ≥99.5%
Glycerine
Formula: HOCH₂CH(OH)CH₂OH
Boiling Pt: 182 °C
Melting Pt: 20 °C
Density: 1.26 g/cm³(20 °C)
Flash Pt: 160 °C
MDL Number: MFCD00004722
CAS Number: 56-81-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3668cce85af1464db21fd098526252076b567956&vid=86a15453b54e7106c121d32e4e716385003c4507&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility in water Fully miscible


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR