Zinc metal, sheets, Length 12", Width 12", Thickness 1/32"

Supplier: AQUA SOLUTIONS MS

Z3388-EA
101223-860EA 101.89 USD
101223-860
Zinc metal, sheets, Length 12", Width 12", Thickness 1/32"
Zinc
Formula: Zn
MW: 65.39 g/mol
Boiling Pt: 908
Melting Pt: 420
MDL Number: MFCD00011291
CAS Number: 7440-66-6
Merck Index: 12,10255

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR