Silver nitrate 0.01 N in 2-propanol (ASTM D3227-13)

Supplier: AQUA SOLUTIONS MS
Danger

8375-4LC 8375-20L 8375-4L 8375-500ML 8375-1L
101433-492EA 201.44 USD
101433-492 101224-852 101224-154 101223-196 101226-340
Silver nitrate 0.01 N in 2-propanol (ASTM D3227-13)
Silver nitrate

Certifications: Traceable to NIST (SRM 919).

Formula: AgNO₃
Melting Pt: –89.5 °C (–129 °F)
Flash Pt: 13 °C (55 °F)
MDL Number: MFCD00003414
CAS Number: 7761-88-8
UN: 1993
ADR: ,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f56d5ff35f4c0937041d9629e42aae69bf1987b3&vid=7ca52a55b378bddc27fb4a51ace3ae27a0451c75&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Color Clear Water White
Form Liquid
Normality 0.01 N
Odor ALCH
Ignition Temperature 425 °C (797 °F)
Density (20 °C) 0.79059 g/cm₃ (6.597 lbs/gal)
Miscibility with Water Fully Miscible


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR