Sodium hydroxide 0.1 M traceable to NIST Standard Reference Material (SRM)

Supplier: AQUA SOLUTIONS MS
Danger

Synonyms: Caustic soda, Caustic soda, Sodium hydrate, Lye, Soda lye

8625-20L 8625-4LC 8625-1L 8625-10L 8625-500ML 8625-4L
101225-214EA 162.59 USD
101225-214 101223-014 101223-036 101223-190 101223-406 101223-140
Sodium hydroxide 0.1 M traceable to NIST Standard Reference Material (SRM)
Sodium hydroxide
Formula: NaOH
MDL Number: MFCD00003548
CAS Number: 1310-73-2

Order Now

Specification Test Results

Normality 0.1000 ± 0.0005 Normal @ 20° C
Appearance and Color Clear Water White Liquid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR