ScienceLine Temp 135 Electrode, YSI
Supplier: YSI
400363
10122-566EA 376.87 USD
10122-566
ScienceLine Temp 135 Electrode, YSI
Temperature Probes
ScienceLine Temp 135, Temperature sensor, Stainless Steel, 120 mm, banana plug, NTC 30 kOhm.
1 m fixed cable.