TruLine ORP 51 Electrode, YSI
Supplier: YSI
400356
10122-564EA 246.09 USD
10122-564
TruLine ORP 51 Electrode, YSI
Electrodes Redox Electrodes
TruLine ORP 51, ORP electrode, Single Junction, Gel electrolyte, Epoxy shaft, 1 m cable, BNC Connector.
Ag/AgCl Reference System. SMEK plug head.