Orthophosphoric acid 0.1 M

Supplier: AQUA SOLUTIONS MS

6849-20L
101203-700EA 270.12 USD
101203-700
Orthophosphoric acid 0.1 M
Orthophosphoric acid
Formula: H₃PO₄
MDL Number: MFCD00011340
CAS Number: 7664-38-2

Order Now

Specification Test Results

Molarity 0.1 M


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR