EVA® Women's Pink Rimless Safety Glasses, PIP
Supplier: PIP
250-10-0920
10120-243CS 2058.93 USD
10120-243
EVA® Women's Pink Rimless Safety Glasses, PIP
Safety Glasses
The first safety eyewear designed specifically for women in the workforce.
  • Smaller and narrower than standard safety glasses; allows the eyewear to better fit the facial structure of a woman
  • Pink rubber co-molded temples and molded nose bridge provide a more secure fit
  • Lightweight, rimless design provides all day user comfort
  • 10 Base curve lens
  • A portion of the proceeds are donated to the American Cancer Society
Clear Lens.
Anti-Scratch Coating
Treated front and back with a clear, scratch-resistant coating for a much harder surface that is more resistant to scratching.
Anti-Fog Coating
Eliminates the condensation of moisture on lenses that causes fogging, ensuring that lenses and vision stay clear.
Certifications: Meets ANSI Z87.1+ standards.