Amersham™ Hybond™ SEQ Western Blotting Membrane, 0.2 PVDF, Cytiva
Supplier: Cytiva
10600030
10120-026PK 867.16 USD
10120-026
Amersham™ Hybond™ SEQ Western Blotting Membrane, 0.2 PVDF, Cytiva
Transfer Membranes
0.2 µm pore size for higher surface area, resulting in better binding of low molecular weight proteins. Amersham Hybond SEQ 0.2 PVDF is a 0.2 µm pore size hydrophobic membrane designed specifically for protein sequencing applications. The small pore size of this membrane minimizes ‘blow-through’ and increases protein binding over a wide range of molecular weights.
  • For Western blotting
  • High protein binding capacity for easy signal detection
  • Chemical resistance needed for N-terminal sequencing
  • High protein retention even after harsh washing steps
  • Efficient capture of proteins during transfer minimizes sample loss
  • Available in popular precut sizes for your application