Human Peripheral Blood CD3 Pan T Cells, Charles River Laboratories Cell Solutions
Supplier: Charles River Laboratories Cell Solutions
PB03C-1 PB03C-2 PB03C-3 PB03NC-1 PB03NC-2 PB03NC-3
101189-998EA 807.69 USD
101189-998 101190-000 101190-002 102830-056 102830-058 102830-060
Human Peripheral Blood CD3 Pan T Cells, Charles River Laboratories Cell Solutions
Blood
Human peripheral blood CD3 Pan T Cells are isolated from peripheral blood mononuclear cells (PBMCs) using either POSITIVE or NEGATIVE immunomagnetic cell separation procedures.
PBMCs are collected in HemaCare's FDA-registered collection centers following cGMP and cGTP collection guidelines from healthy human volunteer donors under IRB approved informed consent by leukapheresis using the COBE® Spectra Apheresis System MNC collection protocol. This mononuclear cell-rich leukapheresis product (LeukoPak) is extracted using continuous flow centrifugal technology directly into the collection bag. Mononuclear cells are purified using density gradient centrifugation techniques.
Serological and Infectious disease testing are performed with each donation. Customers should handle product as if potentially infectious following biosafety level 2 procedures.