SHURDry Slide Dryer II, Triangle Biomedical
Supplier: General Data Healthcare
SD-II-120
10119-900EA 2186.06 USD
10119-900
SHURDry Slide Dryer II, Triangle Biomedical
Slide Warmers/Dryers
SHURDry™ Slide Dryer II is the perfect solution for small volume labs that need a low-volume sliding apparatus that can still dry slides quickly! It offers an uncomplicated, fast method for drying slides to maintain a streamlined histology process
  • See-through glass allows for easy monitoring
  • Perfect solution for small volume labs
  • Easy-to-read indication lights assist with heat control
Monitor heat control utilizing the Amber indication light located by the temperature control, informs the user when the heat is The Red indication light offers protection, and informs the user if overheating occurs. Use the SHURDry™ II with General Data’s extensive line of indispensable products that can be seamlessly integrated into your workspace to increase turnaround time and ensure high-quality results.
The SHURDry™ Slide Dryer II accommodates two slide racks directly from a stainer and allows the slides to be stained in approximately 20 minutes to 1 hour. A see-through glass provides the histologist the visibility needed to examining contents without releasing heat by opening doors and also makes it convenient for cleaning.