Human LeukoPak, Charles River Laboratories Cell Solutions
Supplier: Charles River Laboratories Cell Solutions
PB001F-1 PB001F-2-MNC PB001F-3
101189-944EA 3432.69 USD
101189-944 101189-946 101189-948
Human LeukoPak, Charles River Laboratories Cell Solutions
Blood
Human LeukoPak is collected in HemaCare's FDA-registered collection centers following cGMP and cGTP collection guidelines from IRB consented healthy human volunteer donors by leukapheresis using the COBE® Spectra Apheresis System MNC collection protocol in ACD-A anticoagulant
Serological and Infectious disease testing are performed with each donation. Customers should handle product as if potentially infectious following biosafety level 2 procedures.
A LeukoPak contains ≥85% mononuclear cells