Human Recombinant SFRP4 (from HEK293 Cells)
Supplier: BioVendor
sFRP-4 is a cystein-rich glycoprotein that belongs to the group of G protein-coupled receptor proteins. It is expressed in proliferative endometrium and several types of ovarian, endometrial and brest tumors. It is up-regulated in failing myocardium.
Sequence: PGDYKDDDDK PAGVRGAPCE AVRIPMCRHM PWNITRMPNH LHHSTQENAI LAIEQYEELV DVNCSAVLRF FLCAMYAPIC TLEFLHDPIK PCKSVCQRAR DDCEPLMKMY NHSWPESLAC DELPVYDRGV CISPEAIVTD LPEDVKWIDI TPDMMVQERP LDVDCKRLSP DRCKCKKVKP TLATYLSKNY SYVIHAKIKA VQRSGCNEVT TVVDVKEIFK SSSPIPRTQV PLITNSSCQC PHILPHQDVL IMCYEWRSRM MLLENCLVEK WRDQLSKRSI QWEERLQEQR RTVQDKKKTA GRTSRSNPPK PKGKPPAPKP ASPKKNIKTR SAQKRTNPKR V
Other names: SFRP4
Ordering information: For research use only.
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...