Human Recombinant TPPP (from E. coli)
Supplier: BioVendor
TPPP protein probably participates in the polymerization of tubulin into microtubules. The level of TPPP/p235 in cerebrospinal fluid is significantly higher in multiple sclerosis patients.
Sequence: MKHHHHHHAS MADKAKPAKA ANRTPPKSPG DPSKDRAAKR LSLESEGAGE GAAASPELSA LEEAFRRFAV HGDARATGRE MHGKNWSKLC KDCQVIDGRN VTVTDVDIVF SKIKGKSCRT ITFEQFQEAL EELAKKRFKD KSSEEAVREV HRLIEGKAPI ISGVTKAISS PTVSRLTDTT KFTGSHKERF DPSGKGKGKA GRVDLVDESG YVSGYKHAGT YDQKVQGGK
Other names: TPPP, 25 kDa brain-specific protein, TPPP/p25, p24, p25-alpha, TPPP1
Ordering information: For research use only.
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...