Paraoxonase-1 Human E.coli, 0.1 mg
Supplier: BioVendor
Total 362 AA. MW: 40.68 kDa (calculated). UniProtKB acc.no. P27169 (Met1-Leu355). C-terminal His-tag (7 extra AA). Protein identity confirmed by LC-MS/MS.
- Quality Control Test
- BCA to determine quantity of the protein
- SDS PAGE to determine purity of the protein
- LAL to determine quantity of endotoxin
Sequence: MAKLIALTLL GMGLALFRNH QSSYQTRLNA LREVQPVELP NCNLVKGIET GSEDLEILPN GLAFISSGLK YPGIKSFNPN SPGKILLMDL NEEDPTVLEL GITGSKFDVS SFNPHGISTF TDEDNAMYLL VVNHPDAKST VELFKFQEEE KSLLHLKTIR HKLLPNLNDI VAVGPEHFYG TNDHYFLDPY LQSWEMYLGL AWSYVVYYSP SEVRVVAEGF DFANGINISP DGKYVYIAEL LAHKIHVYEK HANWTLTPLK SLDFNTLVDN ISVDPETGDL WVGCHPNGMK IFFYDSENPP ASEVLRIQNI LTEEPKVTQV YAENGTVLQG STVASVYKGK LLIGTVFHKA LYCELEHHHH HH
Other names: Arylesterase 1, PON 1, Aromatic Esterase 1, A-Esterase 1, K-45, Serum Aryldialkylphosphatase 1
Caution: Research use only.
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...