4-Chloromercuribenzoic acid

Supplier: Pfaltz & Bauer
Danger

C13810-1G
101189-064EA 1115.26 USD
101189-064
4-Chloromercuribenzoic acid
4-Chloromercuriobenzoic acid
Formula: C₇H₅ClHgO₂
Melting Pt: 287 °C
CAS Number: 59-85-8
EINECS: 200-442-6
UN: 2026
ADR: 6.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR