di-Ammonium oxalate monohydrate ≥98%

Supplier: Pfaltz & Bauer
Warning

A30385-100G
101188-522EA 306.14 USD
101188-522
di-Ammonium oxalate monohydrate ≥98%
di-Ammonium oxalate monohydrate
Formula: (NH₄)₂C₂O₄·H₂O
Melting Pt: 133 °C
Density: 1.50 g/cm³
MDL Number: MFCD00149694
CAS Number: 6009-70-7
UN: 3288
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR