Sodium hexametaphosphate

Supplier: Pfaltz & Bauer
Warning

S05625-500G
101187-084EA 247.15 USD
101187-084
Sodium hexametaphosphate
Sodium hexametaphosphate
Formula: (NaO₃P)n
Melting Pt: 628 °C
MDL Number: MFCD00134118
CAS Number: 68915-31-1
EINECS: 272-808-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR