2,2'-Dimethyl-2,2'-azodipropiononitrile ≥98%

Supplier: Pfaltz & Bauer

A37050-25G
101184-722EA 590.65 USD
101184-722
2,2'-Dimethyl-2,2'-azodipropiononitrile ≥98%
2,2'-Dimethyl-2,2'-azodipropiononitrile
Formula: C₈H₁₂N₄
MDL Number: MFCD00013808
CAS Number: 78-67-1
UN: 3234
ADR: 4.1,

Order Now

Specification Test Results

Chemical Name 2,2'-Azobis-2-methylpropionitrile 98%
Synonyms a,a'-Azoisobutyronitrile; AIBN; Azobisisobutyronitrile; VAZO® catalyst 64 free radical source
Molecular Formula (CH₃)₂C(CN)N₂C(CH₃)₂CN
Molecular Weight 164.21
Sensitivity Air and Light sensitive
Storage Temperature 2 to 8 ºC
Appearance White crystalline powder
Melting Point 102-104 ºC
Density 1.1 g/cm³
Stability Stable under normal temperatures and pressures
Incompatibility Aldehydes, Strong oxidizing agents, Alcohols, Alkali metals, Heptane, Contamination, Acetone
Reactivity React with incompatible materials to release other hazardous substances
Conditions to Avoid Heat, flame, sparks, other ignition sources
Hazardous Decomposition Products Carbon oxides, Nitrogen oxides


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR