1,2,4,5-Tetraaminobenzene tetrahydrochloride, Technical Grade

Supplier: Pfaltz & Bauer
Warning

T01680-500MG
101184-311EA 840.72 USD
101184-311
1,2,4,5-Tetraaminobenzene tetrahydrochloride, Technical Grade
1,2,4,5-Tetraaminobenzene tetrahydrochloride
Formula: C₆H₂(NH₂)₄·4HCl
Melting Pt: 300 °C
MDL Number: MFCD00012970
CAS Number: 4506-66-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR