Sodium bitartrate monohydrate ≥98%

Supplier: Pfaltz & Bauer

S04365-100G
101184-128EA 300.12 USD
101184-128
Sodium bitartrate monohydrate ≥98%
Sodium bitartrate monohydrate
Formula: C₄H₅NaO₆·H₂O
Melting Pt: 253 °C
MDL Number: MFCD00150038
CAS Number: 6131-98-2
EINECS: 208-400-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR