Sodium glutamate ≥98%

Supplier: Pfaltz & Bauer
Warning

S05495-500G
101183-584EA 292.28 USD
101183-584
Sodium glutamate ≥98%
Sodium glutamate
Formula: C₅H₈NNaO₄
Melting Pt: 232 °C
MDL Number: MFCD00013074
CAS Number: 142-47-2
EINECS: 205-538-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR