Cupric naphthenate (Cu 6%) in mineral spirits

Supplier: Pfaltz & Bauer

C27410-500G
101181-038EA 210.54 USD
101181-038
Cupric naphthenate (Cu 6%) in mineral spirits
Cupric naphthenate
Formula: Cu(C₁₁H₇O₂)₂
MDL Number: MFCD00146564
CAS Number: 1338-02-9
EINECS: 215-657-0
UN: 1993
ADR: ,

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR