1,3,5-Triethylbenzene ≥98%

Supplier: Pfaltz & Bauer
Warning

T21715-5ML
101180-788EA 291.89 USD
101180-788
1,3,5-Triethylbenzene ≥98%
1,3,5-Triethylbenzene
Formula: C₁₂H₁₈
Boiling Pt: 216 °C
Density: 0.862
Flash Pt: 76 °C
MDL Number: MFCD00009261
CAS Number: 102-25-0
UN: 3082
ADR: 9,III

Order Now

Specification Test Results

Refractive Index 1.495
Storage Temperature 15 - 30 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR