Iodosobenzene ≥95%

Supplier: Pfaltz & Bauer
Danger

I03960-5G
101180-438EA 1063.55 USD
101180-438
Iodosobenzene ≥95%
Iodosobenzene
Formula: C₆H₅IO
Flash Pt: 74 °C
MDL Number: MFCD00039507
CAS Number: 536-80-1
EINECS: 208-648-8
UN: 1479
ADR: 5.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR