Sodium dichromate dihydrate

Supplier: Pfaltz & Bauer

S04313-500G
101180-100EA 646.92 USD
101180-100
Sodium dichromate dihydrate
Sodium dichromate dihydrate
Formula: Na₂Cr₂O₇·2H₂O
MDL Number: MFCD00149166
CAS Number: 7789-12-0
UN: 3087
ADR: 5.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR