Automatic Saybolt and ASTM Colorimeter, Koehler Instrument
Supplier: Koehler
K13150
10118-968EA 41443.18 USD
10118-968
Automatic Saybolt and ASTM Colorimeter, Koehler Instrument
Colorimeters Automatic Transmitance Colorimeters
This high precision spectrophotometer for objective color analysis of petroleum fuels, oils, waxes and petrochemicals according to the Saybolt and ASTM color scales
  • Test results can also be displayed in terms of CIE values and spectral data
  • Routine diagnostic test allows users to conduct periodic checks on the instrument or to identify faults
  • Direct access of the precision filament lamp from outside the instrument allows for easy replacement
  • Equipped with integrated heater unit for melting solid samples such as fats and waxes and preventing from solidification within the cell during testing
The colorimeter is rugged with a fabricated steel housing which is designed to function equally as a QC instrument within the laboratory or on 24 hour operation in a production environment
Certifications: Conforms to ASTM D156, D1500, D6045, and related test specifications.
Delivery information: The colorimeter is supplied with a colored glass filter of known Saybolt value for regular conformance testing.