Core-Type Sampler, Koehler
Supplier: Koehler
K28100
10118-898EA 1587.64 USD
10118-898
Core-Type Sampler, Koehler
Samplers Fluid Samplers
Core-type sampler made of stainless steel and brass construction with simple spring mechanism for taking accurate samples.
  • Obtain bottom samples or samples from any level
  • Butterfly valve on the bottom for ease of sampling
  • Three petcocks for draining at different levels (4, 8 and 12")
Use to manually obtain samples of a liquid, semi-liquid, or solid state whose vapor pressure at ambient conditions is below 101 kPa. Clear polycarbonate sample container ensures to view and determine the amount of sediment and water.
Certifications: Conforms to ASTM D4057.