Autosampler Syringes for Varian ProStar HPLC, Hamilton
Supplier: Hamilton
0159902
10118-432EA 67.36 USD
10118-432
Autosampler Syringes for Varian ProStar HPLC, Hamilton
Syringes Syringes for Chromatography
The Hamilton Syringe is now available for your ProStar AutoSampler
The Star 9100 Autosampler (low volume-low waste) is a stand alone instrument that may be interfaced with a broad range of liquid chromatographs. Up to 105 samples with as many as 8 injections per vial can be accommodated. Single or multiple wash cycles with programmable volumes insure minimal cross-contamination from sample to sample.
Rugged polytetrafluoroethylene (PTFE) plunger tip for GASTIGHT® applications. Economically designed to give true value at an affordable price. Strictly machined tolerances for reproducible results from injection-to-injection. Hamilton glass barrel surface finish for superior syringe life and excellent cost savings. Hamilton offers one quality syringe option for the Varian 9100 HPLC Autosampler.
Certifications: UL Listed. CE compliant.
Ordering information: Needle not included. Accessories not included.