Bacterial Cell Spreaders, Simport Scientific
Supplier: Simport Scientific
L300
10118-328CS 153.52 USD
10118-328
Bacterial Cell Spreaders, Simport Scientific
Spreaders Cell Spreaders
Made of high impact polystyrene, the bacterial cell spreader is designed for easy spreading of cells onto the surface of an agar plate.
  • Sterile
  • No flame sterilization needed
Spreader width is 36 mm.
Packaging: Supplied sterile in individual packs.