Iron(III) citrate monohydrate

Supplier: Pfaltz & Bauer

F00740-100G
101179-890EA 332.94 USD
101179-890
Iron(III) citrate monohydrate
Iron (III) citrate monohydrate
Formula: C₆H₅FeO₇·H₂O
Boiling Pt: 310 °C
Density: 1.762
Storage Temperature: Ambient
MDL Number: MFCD00013098
CAS Number: 2338-05-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR