Anethole cis- and trans-mixture

Supplier: Pfaltz & Bauer
Warning

A32400-100G
101179-858EA 183.86 USD
101179-858
Anethole cis- and trans-mixture
Anethole
Formula: C₁₀H₁₂O
Boiling Pt: 232...235 °C
Melting Pt: 21...23 °C
Density: 0.959
Flash Pt: 88.4 °C
MDL Number: MFCD00009284
CAS Number: 104-46-1
EINECS: 203-205-5

Order Now

Specification Test Results

Refractive Index 1.545


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR