Sodium dodecyl sulfate (SDS) solution 10%, Ultrapure

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Sodium lauryl sulfate, Dodecyl sulfate sodium salt, Lauryl sulfate sodium salt, Dodecyl sulfuric acid sodium salt, Dodecyl sulphate sodium salt, Sodium dodecyl sulfate, Dodecyl sulphuric acid sodium salt, Lauryl sulphate sodium salt, Sodium lauryl sulphate

J77504-K2 J77504-AE J77504-AP
101174-138EA 221.82 USD
101174-138 101171-450 101176-116
Sodium dodecyl sulfate (SDS) solution 10%, Ultrapure
Sodium dodecyl sulphate (SDS)

10% (w/v) solution of sodium dodecyl sulfate (SDS) in high purity dH2O.


  • A(280 nm, 3%, 1 cm): ≤ 0.1
  • A(230 nm, 3%, 1 cm): ≤ 0.2


Solution is 0.2 μm filtered and dispensed into durable, square Nalgene bottles.


Caution: Note: Precipitation of SDS is not unusual. Warm gently to redissolve.

Formula: CH₃(CH₂)₁₁OSO₃Na
Storage Temperature: Ambient
MDL Number: MFCD00036175
CAS Number: 151-21-3
EINECS: 205-788-1
ADR: 4.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fe4cb1741fba41f699a3be006d0b01b1b551898c&vid=c6df9a86fc351b319e68cc79c1c17f733b4fec67&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR