Kanamycin sulfate, Ultrapure

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Kanamycin Monosulfate, (2R,3S,4S,5R,6R)-2-(Aminomethyl)-6-(((1R,2R,3S,4R,6S)-4,6-diamino-3-(((2S,3R,4S,5S,6R)-4-amino-3,5-dihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-2-hydroxycyclohexyl)oxy)tetrahydro-2H-pyran-3,4,5-triol sulfate, Kanamycin A, Kanamycin Monosulphate, (2R,3S,4S,5R,6R)-2-(Aminomethyl)-6-(((1R,2R,3S,4R,6S)-4,6-diamino-3-(((2S,3R,4S,5S,6R)-4-amino-3,5-dihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-2-hydroxycyclohexyl)oxy)tetrahydro-2H-pyran-3,4,5-triol sulphate

J17924-14 J17924-06
101175-072EA 290.35 USD
101175-072 101175-582
Kanamycin sulfate, Ultrapure
Kanamycin sulphate

A selectable marker for cosmid vectors used in transfection of mammalian cells.


It binds to ribosomal components and inhibits protein synthesis. Kanamycin is inactivated by an aminoglycoside phospho transferase (MW 25,000) in the periplasmic space.

Formula: C₁₈H₃₈N₄O₁₅S
MW: 582.6 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00070253
CAS Number: 25389-94-0
EINECS: 246-933-9
Merck Index: 14,5281

Order Now

Specification Test Results

Identity (A, B, C) Passes Test
Potency ≥750 µg of C₁₈H₃₆N₄O₁₁/mg powder
Residue on Ignition ≤1.0%
Impurity Content Organic Impurities Meets the Requirements
Loss on Drying ≤4.0%
pH (1%, H₂O) 6.5 - 8.5


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR