Sodium azide 99+%

Supplier: Thermo Fisher Scientific
Danger

J21610-36 J21610-22 J21610-A1
101171-154EA 179 USD
101171-154 101175-160 103728-110
Sodium azide 99+%
Sodium azide
Formula: NaN₃
Melting Pt: ∼ 275 °C (dec.)
Density: 1.846
Storage Temperature: Ambient
MDL Number: MFCD00003536
CAS Number: 26628-22-8
EINECS: 247-852-1
UN: 1687
ADR: 6.1,II
Merck Index: 14,8581

Order Now

Specification Test Results

Form White crystalline powder
Assay (Unspecified) ≥99.0%
Loss on Drying ≤0.5%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR