γ-(N-Morpholino)propanesulfonic acid (MOPS) ≥98.5% (dry basis), Ultrapure

Supplier: Thermo Fisher Scientific

J19256-A1 J19256-22
101171-042EA 431.17 USD
101171-042 101175-108
γ-(N-Morpholino)propanesulfonic acid (MOPS) ≥98.5% (dry basis), Ultrapure
γ-(N-Morpholino)propanesulphonic acid (MOPS)
Formula: C₇H₁₅NO₄S
Melting Pt: 277…280 °C (dec.)
Flash Pt: 110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00006183
CAS Number: 1132-61-2
EINECS: 214-478-5
Merck Index: 14,6265

Order Now

Specification Test Results

Assay (Dry Basis) ≥98.5%
Assay Absorbance (250 nm, 1 M, 1 cm) ≤0.1
Chloride Content ≤0.01%
Identification (FTIR) Conforms
Residue on Ignition ≤0.1%
Impurity Content Sulphate (SO₄) ≤0.02%
Loss on Drying ≤0.5%
pKa (20 °C) 7.20 ±0.15
Iron (Fe) ≤5 ppm
Lead (Pb) ≤10 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR