Sodium dodecyl sulfate (SDS) ≥95% (dye basis)

Supplier: Thermo Fisher Scientific
Danger

J18220-36 J18220-A7 J18220-A1
101173-766EA 65 USD
101173-766 101170-512 101175-088
Sodium dodecyl sulfate (SDS) ≥95% (dye basis)
Sodium dodecyl sulphate (SDS)
Formula: CH₃(CH₂)₁₁OSO₃Na
Melting Pt: ∼ 206 °C
Storage Temperature: Ambient
MDL Number: MFCD00036175
CAS Number: 151-21-3
EINECS: 205-788-1
ADR: 4.1,II
Merck Index: 14,8636
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fe4cb1741fba41f699a3be006d0b01b1b551898c&vid=c6df9a86fc351b319e68cc79c1c17f733b4fec67&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (Dry Basis) ≥95%
Identification (FTIR) Conforms


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR