EDTA tetrasodium salt dihydrate 99-101%, Ultrapure

Supplier: Thermo Fisher Scientific
Danger

J15700-A1 J15700-C4
101175-024EA 109.54 USD
101175-024 103728-092
EDTA tetrasodium salt dihydrate 99-101%, Ultrapure
EDTA tetrasodium salt dihydrate
Formula: C₁₀H₁₂N₂Na₄O₈·2H₂O
Storage Temperature: Ambient
MDL Number: MFCD00150026
CAS Number: 10378-23-1

Order Now

Specification Test Results

Form White crystalline powder
Assay (unspecified) 99 to 101% (as is)
Identification (FTIR) Conforms
pH (1%, H2O) 10.7 to 11.7
pH (5%, H2O) 10.7 to 12.0


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR