L(+)-Cysteine hydrochloride monohydrate 98.5-101.5% (dry basis)

Supplier: Thermo Fisher Scientific
Warning

J14035-A1 J14035-22 J14035-A7 J14035-14
101174-990EA 396.28 USD
101174-990 101174-992 101171-578 103728-078
L(+)-Cysteine hydrochloride monohydrate 98.5-101.5% (dry basis)
L(+)-Cysteine hydrochloride monohydrate
Formula: HSCH₂CH(NH₂)COOH·HCl·H₂O
Melting Pt: ∼ 175 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00065606
CAS Number: 7048-04-6
EINECS: 200-157-7
Merck Index: 14,2781
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=18703c57a6b5f299a5f38f5d92b333d3d65de497&vid=10395059845247050509d84df67aefcb3c98806c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Form White crystalline powder
Assay (unspecified) 98.5 to 101.5% (dry basis)
Identification (FTIR) Conforms
Residual halide: Chlorine (Cl) 19.8 - 20.8%
Residue on ignition ≤ 0.1%
Impurity content: Sulfate (SO4) ≤ 0.03%
Loss on Drying 8.0 to 12.0%
Optical Rotation +5.7° to 6.8° (c=8 in in 6N HCl, dry basis)
Thin Layer Chromatography Homogeneous
Trace Metal: Arsenic (As) ≤ 1.5 ppm
Trace Metal: Heavy Metals (Pb) ≤ 10 ppm
Trace Metal: Iron (Fe) ≤ 10 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR