Human Recombinant Gastrokine-2 (from E. coli)
Supplier: BioVendor
RD172269100
101174-836EA
583.35
USD
101174-836
Human Recombinant Gastrokine-2 (from E. coli)
Proteins and Peptides
Gastrokine-2 is expressed in gastric mucosa.
Sequence: MKHHHHHHAS YEVFNIISPS NNGGNVQETV TIDNEKNTAI INIHAGSCSS TTIFDYKHGY IASRVLSRRA CFILKMDHQN IPPLNNLQWY IYEKQALDNM FSSKYTWVKY NPLESLIKDV DWFLLGSPIE KLCKHIPLYK GEVVENTHNV GAGGCAKAGL LGILGISICA DIHV
Other names: Blottin, GKN2, BLOT, GDDR, TFIZ1, UNQ465/PRO813
Ordering information: For research use only.
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...