Citric acid, anhydrous 99.5-100.05% (dry basis)

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 2-Hydroxypropane-1,2,3-tricarboxylic acid, Hydroxytricarballylic acid

J13729-36 J13729-A1
101174-286EA 76.89 USD
101174-286 101173-550
Citric acid, anhydrous 99.5-100.05% (dry basis)
Citric acid
Formula: HOC(COOH)(CH₂COOH)₂
MW: 192.13 g/mol
Boiling Pt: 310
Melting Pt: 153…154 °C
Density: 1.665
Storage Temperature: Ambient
MDL Number: MFCD00011669
CAS Number: 77-92-9
EINECS: 201-069-1
REACH: 01-2119457026-42-0000
Merck Index: 14,2326
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d60f5a9b8e6c6dfb7eda04e1a1d5d6d971f92444&vid=caf4f9effc7fd22b942f7f92be770b06e40637a4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99.5 - 100.5% (dry basis)
Assay, Readily Carbonizable Substances Passes Test
Identification (FTIR) Conforms
Residue on Ignition ≤0.1%
Impurity Content, Oxalate Passes Test
Impurity Content, Sulphate (SO₄) Passes test
Water Content (Karl Fischer Titration) ≤1.0%
Heavy Metals (Pb) ≤10 µg/gm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR