Agarose LE, Ultrapure for molecular biology, for electrophoresis

Supplier: Thermo Fisher Scientific

J32802-22 J32802-36 J32802-30 J32802-14 J32802-A1
101174-042EA 284.12 USD
101174-042 101170-230 101175-234 101176-084 103728-122
Agarose LE, Ultrapure for molecular biology, for electrophoresis
Agarose
Melting Pt: 60…90 °C
Storage Temperature: Ambient
MDL Number: MFCD00081294
CAS Number: 9012-36-6
EINECS: 232-731-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=526c20a29610acf6900f898ff245c5602558c785&vid=ae34e4c76a4d7838d4dc9501397f6741b1b97d5e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Form Granular, free-flowing powder
Assay (unspecified) Gel Background Passes test
Assay (unspecified) Gel Point (1.5%) 36 ± 1.5°C
Assay (unspecified) Gel Strength (1%) ≥ 1200 gm/cm2
Assay (unspecified) Gel Strength (1.5%) ≥ 2500 gm/cm2
Comment: EEO (-Mr) 0.05 - 0.13
Comment: Turbidity (1.5%) ≤ 40 Np
DNase Passes test
Residual water ≤ 7%
Residue on ignition ≤ 0.4%
Impurity content: Sulfate (SO4) ≤ 0.14%
Melting Point: Remelt Point (1.5%) 88 ± 1.5°C
RNase Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR