Cholesterol

Supplier: Thermo Fisher Scientific

J13580-22 J13580-06
101172-320EA 768.88 USD
101172-320 101173-546
Cholesterol
Cholesterol
Formula: C₂₇H₄₆O
Boiling Pt: 360 °C
Melting Pt: 147…150 °C
Density: 1.067
Storage Temperature: Refrigerator
MDL Number: MFCD00003646
CAS Number: 57-88-5
EINECS: 200-353-2
Merck Index: 14,2201

Order Now

Specification Test Results

Identification By IR
Thin Layer Chromatography Homogeneous


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR