Agar, Ultrapure for bacteriology

Supplier: Thermo Fisher Scientific

J10906-P2 J10906-P5 J10906-Q1
101170-358EA 150.78 USD
101170-358 101170-360 101173-464
Agar, Ultrapure for bacteriology
Agar
Melting Pt: 90 °C
Storage Temperature: Ambient
MDL Number: MFCD00081288
CAS Number: 9002-18-0
EINECS: 232-658-1
Merck Index: 14,184
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=36877cbcfaca6a0ace648ee20ae51b3467f2748c&vid=4295a0dcbc54bbbd374a5fb934297574c6848d32&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay Gel Point (1.5%) 32 - 38 °C
Assay Gel Strength (1.5%) 550 - 950 gm/cm²
Assay Gel pH (1.5%) after Autoclaving 6.0 - 7.5
Ash ≤6.5%
Residual Water ≤12%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR